Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG7 Antibody (OTI1C1), Allophycocyanin/Cy7, Novus Biologicals™
SDP

Catalog No. N274327APC7 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274327APC7 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274327APC7 Supplier Novus Biologicals Supplier No. NBP274327APCCY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPG7 Monoclonal specifically detects SPG7 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Flow Cytometry.

Specifications

Antigen SPG7
Applications Flow Cytometry
Classification Monoclonal
Clone OTI1C1
Conjugate APC-Cyanine7
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias CARMGC126331, cell matrix adhesion regulator, CMARMGC126332, EC 3.4.24, EC 3.4.24.-, FLJ37308, paraplegin, PGNcell adhesion regulator, spastic paraplegia 7 (pure and complicated autosomal recessive), Spastic paraplegia 7 protein, SPG5C
Gene Symbols SPG7
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 300-573 of human SPG7 (NP_003110) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 6687.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.