Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPG8 Antibody, Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NB183894 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB183894 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB183894 Supplier Novus Biologicals Supplier No. NBP282036AF594
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

SPG8 Polyclonal antibody specifically detects SPG8 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen SPG8
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate Alexa Fluor 594
Formulation 50mM Sodium Borate
Gene Alias KIAA0196, MGC111053, spastic paraplegia 8 (autosomal dominant), SPG8, strumpellin, WASH complex subunit strumpellin
Host Species Rabbit
Immunogen SPG8 antibody was raised against a 19 amino acid synthetic peptide near the carboxy terminus of human SPG8. The immunogen is located within the last 50 amino acids of SPG8. Amino Acid Squence: DYVRYTKLPRRVAEAHVPN
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 9897
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.