Learn More
Eisco™ Sphalerite Mineral Specimen
SCI_ED
Catalog No. S27606
Click to view available options
Quantity:
1/Pk.
12/Pk.
Sphalerite is a sulfide often mined for its zinc content and is typically associated with other sulfide minerals such as pyrite and chalcopyrite.
  • Sphalerite mineral sample
  • Often mined as a zinc ore
  • Perfect size for use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Sphalerite sometimes exhibits triboluminescence, meaning it will glow when fractured
  • Packaged in a clearly labeled, barcode bag for ideal storage
Specifications
Product Type Sulfide Mineral, Sphalerite
Packaging Clearly labeled, barcode bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.