Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sphingosine Kinase 1/SPHK1 Antibody (M540), Novus Biologicals™
SDP

Catalog No. NB387836 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387836 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387836 Supplier Novus Biologicals Supplier No. NBP323419
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Sphingosine Kinase 1/SPHK1 Monoclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Sphingosine Kinase 1/SPHK1
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone M540
Conjugate Unconjugated
Dilution Western Blot 1:250, Immunocytochemistry/ Immunofluorescence 1:200
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1
Host Species Rat
Immunogen Clone M540 was generated from a Sphingosine Kinase 1/SPHK1 synthetic peptide (coupled to carrier) corresponding to amino acids in the C-terminal region of mouse Sphingosine Kinase 1/SPHK1. This sequence has high homology to human and rat Sphingosine Kinase 1/SPHK1, and is not homologous to sequences in Sphingosine Kinase 2/SPHK2. [UniProt# Q8CI15]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8877
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.