Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sphingosine Kinase 1/SPHK1 Antibody (OTI1A6), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. N274332J646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N274332J646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N274332J646 Supplier Novus Biologicals Supplier No. NBP274332JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Sphingosine Kinase 1/SPHK1 Monoclonal specifically detects Sphingosine Kinase 1/SPHK1 in Human, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Sphingosine Kinase 1/SPHK1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A6
Conjugate Janelia Fluor 646
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1
Gene Symbols SPHK1
Host Species Mouse
Immunogen Full length recombinant protein of human Sphingosine Kinase 1/SPHK1 (NP_068807) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8877.0
Target Species Human, Rat, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.