Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sphingosine Kinase 1/SPHK1, p Ser225 Antibody, Novus Biologicals™
SDP

Catalog No. NB387834 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387834 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387834 Supplier Novus Biologicals Supplier No. NBP323421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sphingosine Kinase 1/SPHK1 Polyclonal antibody specifically detects Sphingosine Kinase 1/SPHK1 in Human samples. It is validated for Western Blot

Specifications

Antigen Sphingosine Kinase 1/SPHK1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:1000
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias EC 2.7.1.91, sphingosine kinase 1, SPHKSK 1, SPK, SPK 1
Host Species Rabbit
Immunogen Phospho-Sphingosine Kinase 1/SPHK1 (Ser-225) synthetic peptide (coupled to carrier protein) corresponding to amino acids surrounding serine 225 in human Sphingosine Kinase 1/SPHK1. This sequence has 4 amino acid differences from mouse and 5 from rat Sphingosine Kinase 1/SPHK1, and is not homologous to sequences in SK2. The conserved site in rat and mouse Sphingosine Kinase 1/SPHK1 is at serine 224. [UniProt# Q9NYA1]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 8877
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.