Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sphingosine Kinase 2/SPHK2 Antibody, Novus Biologicals™
SDP

Catalog No. NB387832 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB387832 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB387832 Supplier Novus Biologicals Supplier No. NBP323423
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Sphingosine Kinase 2/SPHK2 Polyclonal antibody specifically detects Sphingosine Kinase 2/SPHK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation

Specifications

Antigen Sphingosine Kinase 2/SPHK2
Applications Western Blot, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:250, Immunocytochemistry/ Immunofluorescence 1:100, Immunoprecipitation 1:100
Formulation PBS, 1 mg/mL BSA and 50% Glycerol
Gene Alias EC 2.7.1.91, SK 2, sphingosine kinase 2, sphingosine kinase type 2 isoform, SPK 2
Host Species Rabbit
Immunogen Sphingosine Kinase 2/SPHK2 (N-terminal region) synthetic peptide (coupled to KLH) corresponding to amino acid residues in the N-terminal region of human Sphingosine Kinase 2/SPHK2. This peptide sequence is highly conserved in rat and mouse Sphingosine Kinase 2/SPHK2 proteins, and has no homology to SK1. [UniProt# Q9NRA0]
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 56848
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.