Learn More
Invitrogen™ SPHK2 Polyclonal Antibody

Description
A suggested positive control for this product is HEK-293 cells.
Specifications
Specifications
| Antigen | SPHK2 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.08% sodium azide |
| Gene | Sphk2 |
| Gene Accession No. | Q9NRA0 |
| Gene Alias | C76851; SK 2; SK2; SK-2; Sphingosine kinase 2; sphingosine kinase type 2; Sphk2; SPK 2; SPK-2; Spk2, Sk2 |
| Gene Symbols | Sphk2 |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.