Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spi-B Antibody, Novus Biologicals™
SDP

Catalog No. NBP170345SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP170345SS 0.025 mg
NBP170345 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP170345SS Supplier Novus Biologicals Supplier No. NBP170345SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Spi-B Polyclonal specifically detects Spi-B in Human, Mouse, Bovine, Canine, Equine, Primate samples. It is validated for Western Blot.

Specifications

Antigen Spi-B
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 3-5 ug/ml
Formulation PBS containing 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. Q01892
Gene Alias SPI-B, Spi-B transcription factor (Spi-1/PU.1 related), transcription factor Spi-B
Gene Symbols SPIB
Host Species Rabbit
Immunogen A portion of amino acid 15-65 of human Spi-B was used as the immunogen.
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6689
Test Specificity Reacts in Chimpanzee, Cow, Dog, Horse, Human, Mouse, New World monkey and Orangutan. Predicted to react in Lizard. The amino acid sequence used as immunogen is 100% homologous in human, new world monkey, chimpanzee, cow, orangutan, rhesus monkey, wild pig, 94% homologous in panda, 89% homologous in dog, horse, 82% homologous in mouse, rat and 52% homologous in lizard.
Target Species Human, Mouse, Bovine, Canine, Equine, Primate
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.