Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPIC Antibody (PCRP-SPIC-2C5), DyLight 755, Novus Biologicals™
SDP

Catalog No. NB381437 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB381437 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB381437 Supplier Novus Biologicals Supplier No. NBP308334IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPIC Monoclonal antibody specifically detects SPIC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Protein Array

Specifications

Antigen SPIC
Applications Flow Cytometry, Immunofluorescence, Protein Array
Classification Monoclonal
Clone PCRP-SPIC-2C5
Conjugate DyLight 755
Formulation 50mM Sodium Borate
Gene Alias MGC40611, SPI-C, Spi-C transcription factor (Spi-1/PU.1 related), transcription factor Spi-C
Host Species Mouse
Immunogen Recombinant full-length human SPIC protein (Uniprot: Q8N5J4)
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 121599
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.