Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPICE1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$158.00 - $487.50
Specifications
| Antigen | SPICE1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP170720UL
![]() |
Novus Biologicals
NBP17071120UL |
20 μL |
Each for $158.00
|
|
|||||
NBP170711
![]() |
Novus Biologicals
NBP170711 |
100 μL |
Each for $487.50
|
|
|||||
Description
SPICE1 Polyclonal specifically detects SPICE1 in Human samples. It is validated for Western Blot.Specifications
| SPICE1 | |
| Polyclonal | |
| Rabbit | |
| CCDC52, FLJ26064, SPICE, spindle and centriole associated protein 1 | |
| SPICE1 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| 152185 | |
| Synthetic peptides corresponding to CCDC52(coiled-coil domain containing 52) The peptide sequence was selected from the N terminal of CCDC52. Peptide sequence TVTDLTVHRATPEDLVRRHEIHKSKNRALVHWELQEKALKRKWRKQKPET. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title