Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spike RBD Antibody (1042425), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB305452 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305452 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305452 Supplier Novus Biologicals Supplier No. FAB10827X
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Spike RBD Monoclonal specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.

Specifications

Antigen Spike RBD
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 1042425
Conjugate Alexa Fluor 532
Dilution Western Blot, Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Chinese Hamster Ovary cell line CHO-derived SARS-CoV Spike RBD, Arg306-Phe527, Accession # NP_0828851.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.