Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spike RBD Antibody (1049349), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB305342 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB305342 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB305342 Supplier Novus Biologicals Supplier No. FAB11055W
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Spike RBD Monoclonal specifically detects Spike RBD in SARS-CoV-2 samples. It is validated for Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Spike RBD
Applications Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 1049349
Conjugate DyLight 650
Dilution Flow Cytometry, Immunocytochemistry
Formulation 50mM Sodium Borate
Host Species Mouse
Immunogen Human embryonic kidney cell HEK293-derived SARS-CoV-2 Spike RBD, Val16-Pro681, Accession # YP_009724390.1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species SARS-CoV-2
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.