Learn More
SPIN90 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$155.13 - $588.46
Specifications
Antigen | SPIN90 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-2616
![]() |
Bethyl Laboratories, Inc
IHC00731T |
10 μL |
Each for $155.13
|
|
|||||
50-157-2615
![]() |
Bethyl Laboratories, Inc
IHC00731 |
100 μL |
Each for $588.46
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Guinea pig_10141, Gorilla, Chimpanzee, Northern white-cheeked gibbon, Chinese hamster and
The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintece of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified.Specifications
SPIN90 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
AF3P21, DIP, DIP1, ORF1, SPIN90, WASLBP, WISH, 54 kDa VacA-interacting protein, 54 kDa vimentin-interacting protein, 90 kDa SH3 protein interacting with Nck, DIP-1, NCK-interacting protein with SH3 domain, SH3 adapter protein SPIN90, SH3 protein interacting with Nck, 90 kDa, VIP54, WASP-interacting SH3-domain | |
NCKIPSD | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human | |
Q9NZQ3 | |
51517 | |
Between 600 and 650 | |
Primary | |
4° C | |
NCKIPSD |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.