Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Spinesin Antibody (OTI4G4), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NP274337A30 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP274337A30 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP274337A30 Supplier Novus Biologicals Supplier No. NBP274337AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Spinesin Monoclonal specifically detects Spinesin in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Spinesin
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI4G4
Conjugate Alexa Fluor 350
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias EC 3.4.21, EC 3.4.21.-, EC 3.4.21.4, MGC141886, MGC148044, Spinesin, transmembrane protease serine 5, transmembrane protease, serine 5
Gene Symbols TMPRSS5
Host Species Mouse
Immunogen Full length recombinant protein of human Spinesin (NP_110397) produced in HEK293T cells.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 80975
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.