Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPINK1 Antibody, PE, Novus Biologicals™
SDP

Catalog No. NB320092 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB320092 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB320092 Supplier Novus Biologicals Supplier No. NBP298602PE
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPINK1 Polyclonal antibody specifically detects SPINK1 in Human samples. It is validated for Immunohistochemistry (Paraffin)

Specifications

Antigen SPINK1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate PE
Dilution Immunohistochemistry-Paraffin
Formulation PBS
Gene Alias pancreatic secretory trypsin inhibitor, PCTTSpink3, PSTISerine protease inhibitor Kazal-type 1, serine peptidase inhibitor, Kazal type 1, serine protease inhibitor, Kazal type 1, TATITumor-associated trypsin inhibitor
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human SPINK1 fragment.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 6690
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.