Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPINK2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB341490 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341490 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341490 Supplier Novus Biologicals Supplier No. NBP282015MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPINK2 Polyclonal antibody specifically detects SPINK2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SPINK2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Acrosin-trypsin inhibitor, Kazal type 2 (acrosin-trypsin inhibitor), serine peptidase inhibitor, Kazal type 2 (acrosin-trypsin inhibitor), serine protease inhibitor Kazal-type 2, serine protease inhibitor, Kazal type, 2 (acrosin-trypsin inhibitor)
Host Species Rabbit
Immunogen SPINK2 antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of mouse SPINK2. The immunogen is located within the last 50 amino acids of SPINK2. Amino Acid Squence: KIREDGSHINIIKDE
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6691
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.