Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPINK4 Antibody (285), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB301773 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB301773 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB301773 Supplier Novus Biologicals Supplier No. NBP289955MFV450
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SPINK4 Monoclonal antibody specifically detects SPINK4 in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen SPINK4
Applications ELISA
Classification Monoclonal
Clone 285
Conjugate mFluor Violet 450 SE
Dilution ELISA, Sandwich ELISA Capture
Formulation 50mM Sodium Borate
Gene Alias gastrointestinal peptide, MGC133107, serine peptidase inhibitor, Kazal type 4, serine protease inhibitor Kazal-type 4, serine protease inhibitor, Kazal type 4
Host Species Rabbit
Immunogen Rabbit immunized with purified, recombinant Human SPINK4 (Accession#: NP_055286.1, Met1-Cys86)
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 27290
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.