Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SPINK4 Recombinant Rabbit Monoclonal Antibody (265), Invitrogen™

Rabbit Recombinant Monoclonal Antibody

Supplier:  Invitrogen™ MA543353

 View more versions of this product

Catalog No. PIMA543353


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

Specificity: Human SPINK4

SPINK4 (Serine Peptidase Inhibitor, Kazal Type 4) is a Protein Coding gene. Gene Ontology (GO) annotations related to this gene include serine-type endopeptidase inhibitor activity.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SPINK4
Recombinant Monoclonal
1 mg/mL
PBS with no preservative
O60575
SPINK4
Recombinant Human SPINK4 Protein.
50 μL
Primary
Human
Antibody
IgG
ELISA
265
Unconjugated
SPINK4
epididymis luminal protein 136; gastrointestinal peptide; HEL136; PEC60; PEC-60; Peptide PEC-60 homolog; serine peptidase inhibitor, Kazal type 4; serine protease inhibitor Kazal-type 4; serine protease inhibitor, Kazal type 4; SPINK4
Rabbit
Protein A
RUO
27290
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.