Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPIRE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 177571AP150UL

Description
Acts as a actin nucleation factor, remains associated with the slow-growing pointed end of the new filament. Involved in vesicle transport processes providing a novel link between actin organization and intracellular transport.Specifications
SPIRE2 | |
Polyclonal | |
Unconjugated | |
SPIRE2 | |
KIAA1832, Protein spire homolog 2, Spir 2, SPIR2, spire homolog 2 (Drosophila), SPIRE2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
234857, 307925, 84501 | |
-20°C | |
Liquid |
Western Blot | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8K1S6, Q8WWL2 | |
SPIRE2 | |
SPIRE2 Fusion Protein Ag12117 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction