Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPIRE2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | SPIRE2 |
---|---|
Concentration | 0.35 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-1769
![]() ![]() |
Proteintech Group Inc
177571AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Acts as a actin nucleation factor, remains associated with the slow-growing pointed end of the new filament. Involved in vesicle transport processes providing a novel link between actin organization and intracellular transport.Specifications
SPIRE2 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8K1S6, Q8WWL2 | |
234857, 307925, 84501 | |
SPIRE2 Fusion Protein Ag12117 | |
Primary | |
-20°C | |
SPIRE2 |
0.35 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
KIAA1832, Protein spire homolog 2, Spir 2, SPIR2, spire homolog 2 (Drosophila), SPIRE2 | |
SPIRE2 | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title