Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Splicing Factor 1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15732220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15732220 20 μL
NBP157322 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15732220 Supplier Novus Biologicals Supplier No. NBP15732220UL

Rabbit Polyclonal Antibody

Splicing Factor 1 Polyclonal specifically detects Splicing Factor 1 in Human samples. It is validated for Western Blot.

Specifications

Antigen Splicing Factor 1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.2-1 ug/ml
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q15637
Gene Alias Mammalian branch point-binding protein, mBBP, splicing factor 1, Transcription factor ZFM1, ZFM1BBP, Zinc finger gene in MEN1 locus, Zinc finger protein 162D11S636, ZNF162MBBP
Gene Symbols SF1
Host Species Rabbit
Immunogen Synthetic peptides corresponding to SF1(splicing factor 1) The peptide sequence was selected from the C terminal of SF1 (NP_004621). Peptide sequence MASSTPLPWQQNTTTTTTSAGTGSIPPWQQQQAAAAASPGAPQMQGNPTM.
Molecular Weight of Antigen 68 kDa
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7536
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.