Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Splicing Factor 1 Antibody (OTI7H9), DyLight 405, Novus Biologicals™
SDP

Catalog No. NBP274338V Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP274338V 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP274338V Supplier Novus Biologicals Supplier No. NBP274338V
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Splicing Factor 1 Monoclonal specifically detects Splicing Factor 1 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen Splicing Factor 1
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI7H9
Conjugate DyLight 405
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Mammalian branch point-binding protein, mBBP, splicing factor 1, Transcription factor ZFM1, ZFM1BBP, Zinc finger gene in MEN1 locus, Zinc finger protein 162D11S636, ZNF162MBBP
Gene Symbols SF1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-276 of human Splicing Factor 1 (NP_973727) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 7536.0
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.