Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Restek™ Split Liners for Agilent™/Finnigan™ GCs: Cup Splitter for Siltek™ Liner
SDP

Catalog No. 06717814
Click to view available options
Includes:
Siltek Cup Splitter
Siltek Cup Splitter w/CarboFrit
Siltek Cup Splitter w/Wool

For high or low molecular weight compounds

  • Increased sample residence time in the liner for homogeneous vaporization
  • 4mm I.D.
  • With Siltek™ deactivation
  • Available with or without packing

Specifications

Length (Metric) 7.85 cm
Product Type Siltek Cup Splitter Inlet Liner
Quantity 5/Pk.
Includes Siltek Cup Splitter
Diameter (Metric) Inner 0.4 cm
Diameter (Metric) Outer 0.63 cm
For Use With (Equipment) Agilent GCs Equipped with Split/Splitless Inlets
Liner Material Borosilicate Glass
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.