Learn More
Invitrogen™ SPOP Recombinant Rabbit Monoclonal Antibody (2N8L8)

Description
Positive Samples: PC-3 Immunogen sequence: VFKEMMCFIY TGKAPNLDKM ADDLLAAADK YALERLKVMC EDALCSNLSV ENAAEILILA DLHSADQLKT QAVDFINYHA SDVLETSGWK SMVVSHPHLV A.
Specifications
Specifications
| Antigen | SPOP |
| Applications | ELISA, Western Blot |
| Classification | Recombinant Monoclonal |
| Clone | 2N8L8 |
| Concentration | 1.08 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | SPOP |
| Gene Accession No. | O43791 |
| Gene Alias | AI315626; BTBD32; HIB homolog 1; Pcif1; PDX-1 C-terminal-interacting factor 1; roadkill homolog 1; similar to speckle-type POZ protein; speckle type BTB/POZ protein; speckle type POZ protein; speckle-type POZ protein; Spop; SPOP1; TEF2; zgc:64140 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.