Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRED1 Antibody, mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB341837 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB341837 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB341837 Supplier Novus Biologicals Supplier No. NBP281927MFV610
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRED1 Polyclonal antibody specifically detects SPRED1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SPRED1
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias EVH1/Sprouty domain containing protein, FLJ33903, hSpred1, NFLS, spred-1, sprouty-related, EVH1 domain containing 1, sprouty-related, EVH1 domain-containing protein 1, suppressor of Ras/MAPK activation
Host Species Rabbit
Immunogen SPRED1 antibody was raised against a 14 amino acid synthetic peptide near the center of the human SPRED1. The immunogen is located within amino acids 250 - 300 of SPRED1. Amino Acid Squence: RHPDMWKNDLERDD
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 161742
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.