Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRED3 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB396512 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396512 0.025 mg
NBP241091 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB396512 Supplier Novus Biologicals Supplier No. NBP2410910.025MG
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRED3 Polyclonal specifically detects SPRED3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SPRED3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% Sodium Azide
Gene Alias Eve-3, spred-3, Sprouty-Related, EVH1 Domain Containing 3, Sprouty-Related, EVH1 Domain-Containing Protein 3
Gene Symbols SPRED3
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of the human Spred3. The immunogen is located within amino acids 140 - 190 of Spred3. Amino Acid Squence: SHSRQETPPSAAAAPI
Molecular Weight of Antigen 45 kDa
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 399473
Test Specificity This Spred3 antibody is predicted to have no cross-reactivity to Spred1 or Spred2.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.