Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRED3 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB344178 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344178 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344178 Supplier Novus Biologicals Supplier No. NBP241091MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRED3 Polyclonal antibody specifically detects SPRED3 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen SPRED3
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias Eve-3, spred-3, Sprouty-Related, EVH1 Domain Containing 3, Sprouty-Related, EVH1 Domain-Containing Protein 3
Host Species Rabbit
Immunogen Antibody was raised against a 16 amino acid synthetic peptide near the center of the human Spred3. The immunogen is located within amino acids 140 - 190 of Spred3. Amino Acid Squence: SHSRQETPPSAAAAPI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 399473
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.