Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRY1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB324992 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB324992 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB324992 Supplier Novus Biologicals Supplier No. NBP299516MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRY1 Polyclonal antibody specifically detects SPRY1 in Human samples. It is validated for Western Blot

Specifications

Antigen SPRY1
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias Drosophila, homolog of, 1 (antagonist of FGF signaling), sprouty homolog 1, antagonist of FGF signaling (Drosophila), spry-1
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human SPRY1.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Growth and Development, Neuronal Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10252
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.