Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPRY2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP213158 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP213158 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP213158 Supplier Novus Biologicals Supplier No. NBP213158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPRY2 Polyclonal specifically detects SPRY2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen SPRY2
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.04 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1:500-1:5000, ELISA 1:100-1:2000, Immunohistochemistry 10 ug/ml, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. O43597
Gene Alias hSPRY2, MGC23039, protein sprouty homolog 2, sprouty (Drosophila) homolog 2, sprouty 2, sprouty homolog 2 (Drosophila), spry-2
Gene Symbols SPRY2
Host Species Rabbit
Immunogen SPRY2 Antibody was purified from monospecific rabbit antiserum prepared via repeated immunizations with peptide corresponding to an internal region of human sprouty-2. (Uniprot: O43597)
Molecular Weight of Antigen 34.69 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Growth and Development, Neuronal Cell Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 10253
Target Species Human, Monkey, Orangutan
Content And Storage Store at -20°C short term. Aliquot and store at -80°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.