Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPSB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP174266
Description
SPSB2 Polyclonal specifically detects SPSB2 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
SPSB2 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ17395, Gene-rich cluster protein C9, GRCC9MGC2519, splA/ryanodine receptor domain and SOCS box containing 2, SPRY domain-containing SOCS box protein SSB-2, SSB-2, SSB2SPRY domain-containing SOCS box protein 2 | |
Rabbit | |
28 kDa | |
100 μL | |
Signal Transduction | |
84727 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/ml | |
Western Blot 1.0 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Q99619 | |
SPSB2 | |
Synthetic peptides corresponding to the C terminal of SPSB2. Immunizing peptide sequence IGRGKLYHQSKGPGAPQYPAGTQGEQLEVPERLLVVLDMEEGTLGYAIGG. | |
Affinity purified | |
RUO | |
Primary | |
Expected identity based on immunogen sequence: Rat: 86%; Mouse: 86%. | |
Human, Monkey, Rat, Pig, Bovine, Canine, Guinea Pig, Rabbit, Rhesus Macaque | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction