Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SPSB2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$499.50
Specifications
Antigen | SPSB2 |
---|---|
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Regulatory Status | RUO |
Description
SPSB2 Polyclonal specifically detects SPSB2 in Human, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin).Specifications
SPSB2 | |
Unconjugated | |
RUO | |
Q99619 | |
84727 | |
Synthetic peptides corresponding to the C terminal of SPSB2. Immunizing peptide sequence IGRGKLYHQSKGPGAPQYPAGTQGEQLEVPERLLVVLDMEEGTLGYAIGG. | |
Primary |
Polyclonal | |
Rabbit | |
Signal Transduction | |
FLJ17395, Gene-rich cluster protein C9, GRCC9MGC2519, splA/ryanodine receptor domain and SOCS box containing 2, SPRY domain-containing SOCS box protein SSB-2, SSB-2, SSB2SPRY domain-containing SOCS box protein 2 | |
SPSB2 | |
IgG | |
28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title