Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTBN4 Antibody (S393-2), DyLight 550, Novus Biologicals™
SDP

Catalog No. NBP259321R Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259321R 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259321R Supplier Novus Biologicals Supplier No. NBP259321R
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SPTBN4 Monoclonal specifically detects SPTBN4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SPTBN4
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone S393-2
Conjugate DyLight 550
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias beta-IV spectrin, KIAA1642, QV, spectrin beta chain, brain 3, spectrin, beta, non-erythrocytic 4, spectrin, non-erythroid beta chain 3, SPNB4, SPTBN3
Gene Symbols SPTBN4
Host Species Mouse
Immunogen Fusion protein amino acids 1621-1832 (C-terminal repeats 14 to 15) of human Beta4-spectrin
Purification Method Protein G purified
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 57731
Test Specificity Detects >200kDa. Does not cross react with other Beta-spectrins
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.