Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SPTLC3 Antibody, Novus Biologicals™
SDP

Catalog No. NBP233418 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP233418 0.1 mL
NB397431 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP233418 Supplier Novus Biologicals Supplier No. NBP233418
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SPTLC3 Polyclonal specifically detects SPTLC3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen SPTLC3
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100 - 1:250, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
Gene Accession No. Q9NUV7
Gene Alias C20orf38, Chromosome 20 Open Reading Frame 38, dJ718P11, EC 2.3.1.50, hLCB2b, LCB 3, LCB2B, LCB3, Long Chain Base Biosynthesis Protein 2b, Long Chain Base Biosynthesis Protein 3, Serine Palmitoyltransferase 3, Serine Palmitoyltransferase, Long Chain Base Subunit 2-Like (Aminotransferase 2), Serine Palmitoyltransferase, Long Chain Base Subunit 3, Serine-Palmitoyl-CoA Transferase 3, SPT 3, SPT3, SPTLC2L
Gene Symbols SPTLC3
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to amino acids: SYNFLGLAAKYDESMRTIKDVLEVYGTGVASTRHEMGTLDKHKELEDLVAKFLNVEAAMVF
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55304
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.