Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SpUlp1 Antibody, Novus Biologicals™
SDP

Catalog No. NB019360 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB019360 0.025 mg
NBP223621 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB019360 Supplier Novus Biologicals Supplier No. NBP223621SS

Rabbit Polyclonal Antibody

SpUlp1 Polyclonal specifically detects SpUlp1 in Yeast samples. It is validated for Western Blot.

Specifications

Antigen SpUlp1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500-1-1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Anti-SpUlp1, S. pombe Ubiquitin-like Protein 1, SpUlp 1, SpUlp -1, SpUlp1
Host Species Rabbit
Immunogen Reacts with residues 7-20 of the SpUlp1 protein.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Yeast
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.