Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squalane, NF, 97-102%, Spectrum™ Chemical
SDP

Catalog No. 18606983 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
100 mL
500 mL
12 kg
20 L
Packaging:
Amber Glass Bottle

Catalog No. 18-606-983

Supplier: Spectrum Chemical Manufacturing Corporation S198720LTBL

Only null left
Add to Cart
Add to Cart
Request a Quote

S1987, 111-01-3, C30H62

Spectrum™ Chemical Squalane, NF is used as an emollient and moisturizer. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 111-01-3
Molecular Formula C30H62
Molecular Weight (g/mol) 422.83
InChI Key PRAKJMSDJKAYCZ-UHFFFAOYNA-N
IUPAC Name 2,6,10,15,19,23-hexamethyltetracosane
SMILES CC(C)CCCC(C)CCCC(C)CCCCC(C)CCCC(C)CCCC(C)C

Specifications

CAS Max % 1
Boiling Point 212°C
Ignition Residue 0.005
Packaging Amber Glass Bottle
Refractive Index 1.4510-1.4525
Quantity 20 L
Saponification Value 2
Specific Gravity 0.807-0.810
Percent Purity 97 to 102%
Grade NF
Physical Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.