Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Squalane, Spectrum™ Chemical
SDP

Catalog No. 18621476 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
100 mL
20 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-621-476 100 mL
18-621-478 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-621-476 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. S2018100ML
Only null left
Add to Cart
Add to Cart

Chemical Identifiers

CAS 111-01-3
Molecular Weight (g/mol) 422.81 g/mol

Specifications

Melting Point -38°C
Quantity 100 mL
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Squalane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.