Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ergodyne™ Squids™ 3101 Stainless Single Carabiner

Tool lanyard with single stainless steel manual locking screw gate carabiner for a secure connection in more corrosive environments.

$31.50

Specifications

Load Bearing Capacity (English) 15 lb.
Load Bearing Capacity (Metric) 6.8 kg
View More Specs

Products 2
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
19-820-297
View Documents
Ergodyne™
19017
Extended
Each for $31.50
Only null left
Add to Cart
 
19-820-295
View Documents
Ergodyne™
19007
Standard N/A N/A N/A
Description

Description

  • Energy absorbing cord with tubular nylon webbing
  • Manual locking 304 stainless steel carabiners
  • Captive eye carabiner
  • Reflective accent for higher visibility
  • Lock stitching at both ends for extra security
  • Certified to a 2:1 safety factor
Specifications

Specifications

15 lb.
6.8 kg
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.