Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SR 12813
GREENER_CHOICESDP

Catalog No. 296950 Shop All Novus Products
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-695-0 50 mg
29-691-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 29-695-0 Supplier R&D Systems™ Supplier No. 2969/50

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Pregnane X receptor agonist

SR 12813 is a pregnane X receptor (PXR) agonist (EC50 values are 200 and 700 nM for human and rabbit PXR respectively). Activates the farnesoid X receptor (FXR) at μM concentrations. Displays hypocholesterolemic activity via enhanced degradation of HMG-CoA reductase.

Chemical Identifiers

CAS 126411-39-0
Molecular Formula C24H42O7P2

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 504.53
Purity >99%
Molecular Weight (g/mol) 504.541
Chemical Name or Material SR 12813
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.