CAS RN 126411-39-0
IUPAC Name:
diethyl [2-(3,5-di-tert-butyl-4-hydroxyphenyl)-1-(diethoxyphosphoryl)ethenyl]phosphonate
Synonyms:
diethyl 2-(3,5-di-tert-butyl-4-hydroxyphenyl)-1-(diethoxyphosphoryl)ethenylphosphonate
Molecular Weight (g/mol):
504.54
Molecular Formula:
C24H42O7P2
InChi Key:
YQLJDECYQDRSBI-UHFFFAOYSA-N
SMILES:
CCOP(=O)(OCC)C(=CC1=CC(=C(O)C(=C1)C(C)(C)C)C(C)(C)C)P(=O)(OCC)OCC
1
–
2
of
2
results
1
Thermo Scientific Chemicals SR-12813, 98%
CAS: 126411-39-0 Molecular Formula: C24H42O7P2 Synonym: Tetraethyl 2-(3,5-di-tert-butyl-4-hydroxyphenyl)ethenyl-1,1-bisphosphonate;
2
R&D Systems™ SR 12813
CAS: 126411-39-0 Molecular Formula: C24H42O7P2