Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ SR 140333
GREENER_CHOICESDP

Catalog No. 401210 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
40-1210 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 40-1210 Supplier R&D Systems™ Supplier No. 4012/10

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent NK1 receptor antagonist

Potent NK1 receptor antagonist (Ki = 0.74 nM. IC50 = 1.6 nM). Inhibits substance P -invoked calcium mobilization and outward current (IC50 = 1.3 nM); blocks NK1-mediated nitric oxide-dependent vasodilation in vivo.

Chemical Identifiers

CAS 153050-21-6
Molecular Formula C37H45Cl3N2O2

Specifications

Quantity 10 mg
Product Line Tocris
Formula Weight 656.12
Purity >97%
Molecular Weight (g/mol) 656.129
Chemical Name or Material SR 140333
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.