Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 142948
GREENER_CHOICESDP

Catalog No. 23091 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mg
10 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-091 1 mg
23-091-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 23-091 Supplier Tocris Bioscience™ Supplier No. 2309/1
Add to Cart
Add to Cart
This item is not returnable. View return policy

Highly potent NTS receptor antagonist

Potent non-peptide neurotensin (NT) receptor antagonist that binds with high affinity (IC50 = 0.32 - 3.96 nM; Ki < 10 nM). Attenuates amphetamine-induced hyperactivity and is orally active in vivo.

Chemical Identifiers

CAS 184162-64-9
Molecular Formula C39H51N5O6

Specifications

Quantity 1 mg
Formula Weight 685.86
Purity >97%
Molecular Weight (g/mol) 685.866
Chemical Name or Material SR 142948
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.