Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SR 16832

Click to view available options
Quantity:
5 mg
25 mg
Description
SR 16832 is a dual site PPARγ inhibitor. Acts at orthosteric and allosteric sites in the ligand binding domain. Inhibits binding of endogenous ligands and transcriptional activity of PPARγ, more effectively than the orthosteric covalent antagonists GW 9662 (Cat. No. 1508) and T 0070907 (Cat. No. 2301).
Specifications
Specifications
CAS | 2088135-12-8 |
Synonym | 2-Chloro-N-(6-methoxy-4-quinolinyl)-5-nitrobenzamide |
Quantity | 5 mg |
Molecular Formula | C17H12ClN3O4 |
Formula Weight | 357.75 |
Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction