Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ SR 3677 dihydrochloride
GREENER_CHOICE

Catalog No. p-5546039
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent, selective Rho-kinase (ROCK) inhibitor

SR 3677 dihydrochloride is a potent and selective Rho-kinase inhibitor (IC50 values are 3 and 56 nM for ROCK-II and ROCK-I respectively).

Specifications

Quantity 10 mg
Product Type SR 3677 dihydrochloride
Purity >98%
Molecular Weight (g/mol) 481.37
Inhibitors SR 3677 dihydrochloride

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.