Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ SR 7826

Click to view available options
Quantity:
10 mg
50 mg
Description
SR 7826 is a potent and selective LIMK inhibitor (IC50 values are 43, 5536 and 6565 nM for LIMK1, ROCKI and ROCKII, respectively). Inhibits cofilin phosphorylation in A7r5 cells and suppresses migration and invasion of PC-3 cells in vitro. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | N-(2-Hydroxyethyl)-N′-[4,5-methyl-7H-pyrrolo[2,3-d]pyrimidine-4-yl)phenyl]-N-phenylurea |
| CAS | 1219728-20-7 |
| Quantity | 50 mg |
| Target | LIMK Inhibitors |
| Molecular Formula | C22H21N5O2 |
| Formula Weight | 387.43 |
| Purity | >98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction