Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR-AI/MSR Antibody (005), PE, Novus Biologicals™
SDP

Catalog No. NB307409 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307409 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307409 Supplier Novus Biologicals Supplier No. NBP289491PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

SR-AI/MSR Monoclonal antibody specifically detects SR-AI/MSR in Human samples. It is validated for ELISA, ELISA

Specifications

Antigen SR-AI/MSR
Applications ELISA, Sandwich ELISA
Classification Monoclonal
Clone 005
Conjugate PE
Dilution ELISA, Sandwich ELISA Detection
Formulation PBS
Gene Alias CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human SR-AI/MSR (Accession#: NP_619729.1; Lys 77-Leu 451).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4481
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.