Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR-AI/MSR Antibody (351615), PE/Cy7, Novus Biologicals™
SDP

Catalog No. NB313317 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB313317 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB313317 Supplier Novus Biologicals Supplier No. FAB2708PECY7
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SR-AI/MSR Monoclonal specifically detects SR-AI/MSR in Human samples. It is validated for Flow Cytometry.

Specifications

Antigen SR-AI/MSR
Applications Flow Cytometry
Classification Monoclonal
Clone 351615
Conjugate PE-Cyanine7
Dilution Flow Cytometry
Formulation PBS
Gene Alias CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human SR-AI/MSR, Lys77-Leu451, Accession # P21757
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4481
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.