Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR-AI/MSR Antibody, Alexa Fluor™ 647, Novus Biologicals™
SDP

Catalog No. NBP100093Y Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP100093Y 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP100093Y Supplier Novus Biologicals Supplier No. NBP100093AF647
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SR-AI/MSR Polyclonal specifically detects SR-AI/MSR in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen SR-AI/MSR
Applications Western Blot, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Conjugate Alexa Fluor 647
Dilution Western Blot, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P21757
Gene Alias CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A
Gene Symbols MSR1
Host Species Rabbit
Immunogen Synthetic peptide made to a cyclized peptide from human Macrophage Scavenger Receptor I (within residues 400-450). [Swiss-Prot# P21757]
Molecular Weight of Antigen 50 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Adaptive Immunity, Cancer, Immunology, Innate Immunity, Lipid and Metabolism, Neuroscience, Prostate Cancer
Primary or Secondary Primary
Gene ID (Entrez) 4481
Test Specificity Reacts with human and mouse. 100% sequence identity with horse and dog, 93% identity with monkey and bovine, 92% identity with rat.
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.