Learn More
Description
Specifications
Specifications
| Antigen | SR-AI/MSR |
| Applications | Western Blot, Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Concentration | 0.97 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL, Immunocytochemistry/Immunofluorescence 1:50 - 1:200 |
| Formulation | Tris-Glycine, 0.15 M NaCl with 0.05% Sodium Azide |
| Gene Accession No. | P21757 |
| Gene Alias | CD204, CD204 antigen, Macrophage acetylated LDL receptor I and II, macrophage scavenger receptor 1, macrophage scavenger receptor type III, phSR1, phSR2, SCARA1macrophage scavenger receptor types I and II, Scavenger receptor class A member 1, scavenger receptor class A, member 1, SR-A |
| Gene Symbols | MSR1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
