Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SR-BI Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB400101 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB400101 0.1 mL
NB400101SS 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB400101 Supplier Novus Biologicals Supplier No. NB400101
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 98 publications

SR-BI Polyclonal specifically detects SR-BI in Human, Mouse, Rat, Bovine, Chinese Hamster, Mustelid, Primate, S. japonicum samples. It is validated for Western Blot, Simple Western, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting, Flow (Intracellular), Knockout Validated, Block/Neutralize.

Specifications

Antigen SR-BI
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:1000-1:5000, Simple Western 1:100, Flow Cytometry 1:400. Use reported in scientific literature (PMID 23029167), Immunohistochemistry 2.5-5 ug/ml, Immunocytochemistry/Immunofluorescence 1:50-1:1000, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 2.5-5 ug/ml, Immunohistochemistry-Frozen reported in scientific literature (PMID 24244566), Immunoblotting reported in scientific literature (PMID 27599291), Flow (Intracellular), Knockout Validated, Block/Neutralize reported in scientific literature (PMID 12119305; 26905520)
Formulation PBS with 0.02% Sodium Azide
Gene Accession No. Q8WTV0, Q61009, P97943
Gene Alias CD36 and LIMPII analogous 1, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 1, CD36L1scavenger receptor class B type 1, CLA1 CD36 antigen-like 1, CLA-1 HDLQTL6, Collagen type I receptor, thrombospondin receptor-like 1, MGC138242, SCARB1, scavenger receptor class B type III, scavenger receptor class B, member 1, SR-B1, SRB1 scavenger receptor class B member 1, SRBI
Gene Symbols SCARB1
Host Species Rabbit
Immunogen A C-terminal peptide containing residues from mouse SR-BI (within residues 450-509). [Uniprot: Q61009]
Molecular Weight of Antigen 82 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Breast Cancer, Cancer, Cardiovascular Biology, Cholesterol Metabolism, Lipid and Metabolism, Signal Transduction, Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 949
Test Specificity Human, mouse, mink, primate, rat, Chinese Hamster and Schistosoma japonicum. bovine reactivity reported in scientific literature (PMID: 24196350).
Target Species Human, Mouse, Rat, Bovine, Chinese Hamster, Mustelid, Primate, S. japonicum
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.